Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sorvall™ Microtube Adapters

Catalog No. 76003759 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
0.2mL microtubes
For 0.25/0.4mL microtubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
76-003-759 For 0.25/0.4mL microtubes
76-003-750 0.2mL microtubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 76-003-759 Supplier Thermo Scientific™ Supplier No. 76003759
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Accommodate tubes from 0.2 to 0.6mL using Thermo Scientific™ Sorvall™ adapters.

Accommodate tubes from 0.2 to 0.6mL using Thermo Scientific™ Sorvall™ Microtube Adapters.

Ordering Information:

Sold in packs of 24

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) For 0.25/0.4mL microtubes
Description For 0.25/0.4mL microtubes; Red
Color Red
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.