Learn More
Thermo Scientific™ Sorvall ST1 Plus-MD, 120-240V TX-400 Blood Tube Package
Catalog No. 75016007PM Shop All Thermo Scientific Products
Change view
Click to view available options
Sorvall ST1 Plus-MD
Catalog No.
75-016-007PM Sorvall ST1 Plus-MD
1 options
This item is not returnable. View return policy
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. 75-016-007PM Supplier Thermo Scientific™ Supplier No. 75016007PR
This item is not returnable. View return policy
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Waste no time finding the right part numbers for your application! The application driven centrifuge package ensures convenient, fast ordering of all components necessary to run a specific protocol.
Easy to Use:
  • Intuitive controls simplify operation and provide detailed information on a wide range of operating parameters and processing status for optimal performance.
  • Modern industrial design supports continuous improvement in rotor loading ergonomics, user safety and current needed certifications
  • Motorized lid latch enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches.
Thermo Scientific™ Auto-Lock™ III Rotor System:
  • Secure locking system allows easy push-button installation and exchange of rotors.
  • System saves time when switching between applications.
  • Easy access to rotor chamber offers quick cleaning, a healthier working environment and longer unit life.
Thermo Scientific™ ClickSeal™ Bucket Sealing System:
  • Biocontainment solution seals with a snap; eliminates screw caps and complicated clips.
  • System has glove-friendly, one-handed open/close capability.
  • System is certified by CAMR™ in Porton Down, UK.
Thermo Scientific™ SMARTSpin™ Technology:
  • Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option:
  • Technology optimizes safety and improves separations.
Space-Saving Design:
  • Compact dimensions optimize work area and ergonomic height and curved design ensure easy sample loading and unloading and easy cleaning.
Energy Efficient:
  • System offers up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing.
TRUSTED_SUSTAINABILITY
Specifications
Drive System Direct, Brushless induction low profile motor with choices of 9 acceleration, 10 deceleration rates
Green Features Less waste, Energy efficient
Includes Part Numbers In Pk. 75009240 (centrifuge),75003181(rotor),75003655(set of buckets),75003656(lids),75003680(set of adapters),75003681(set of adapters)
Product Type General Purpose Centrifuge Series