Learn More
Thermo Scientific™ Sorvall ST8 Ventilated Package with TX-150 and HighConic Rotor Package
Catalog No. 75200709 Shop All Thermo Scientific Products
Change view
Click to view available options
Includes:
Sorvall ST8 ventilated centrifuge, TX-150 Swinging bucket rotor, TX-150 round buckets, TX-150 ClickSeal biocontainment lids, HighConic rotor
Catalog No. Includes
75-200-709 Sorvall ST8 ventilated centrifuge, TX-150 Swinging bucket rotor, TX-150 round buckets, TX-150 ClickSeal biocontainment lids, HighConic rotor
1 options
Catalog No. 75-200-709 Supplier Thermo Scientific™ Supplier No. 75200709
Thermo Scientific™ Sorvall™ ST 8 small benchtop TX150 and HighConic rotor centrifuge package provides exceptional capacity in a compact design with a smart, simple interface and the flexibility to support numerous fixed angle and swinging bucket rotor applications.
  • Secure, push-button Thermo Scientific™ Auto-Lock™ rotor exchange in as little as three seconds delivers trouble-free rotor installation and removal and clear chamber access for cleaning convenience
  • Biocontainment sealing options, including certified Thermo Scientific™ ClickSeal™ lids offer glove-friendly, one-handed operation
  • Fixed angle flexibility to spin 50mL conical tubes at over 12,000 x g
  • Choose adapters that work best for your application (adapters sold separately)
  • TX-150 swinging bucket rotor spins up to 4 x 50 mL conical tubes, 8 x 15 mL conical tubes and 24 x 5/7 mL blood tubes with biocontainment
  • Package includes:
    • Sorvall ST8 ventilated centrifuge
    • TX-150 Swinging bucket rotor
    • TX-150 round buckets
    • TX-150 ClickSeal biocontainment lids
    • HighConic rotor
TRUSTED_SUSTAINABILITY
Specifications
Includes Sorvall ST8 ventilated centrifuge, TX-150 Swinging bucket rotor, TX-150 round buckets, TX-150 ClickSeal biocontainment lids, HighConic rotor