Learn More
Thermo Scientific™ Sorvall X1 R Pro, 120V TX-400 Cell Culture Package
GSA_VAGREENER_CHOICE
Catalog No. 75016012 Shop All Thermo Scientific Products
This item is not returnable. View return policy
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. 75-016-012 Supplier Thermo Scientific™ Supplier No. 75016012
  • $18,250.00
    $9,705.90 / Each of 1
    Save $8,544.10
    47% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart
Request a Quote
This item is not returnable. View return policy
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Waste no time finding the right part numbers for your application! The application driven centrifuge package ensures convenient, fast ordering of all components necessary to run a specific protocol.
Waste no time finding the right part numbers for your application! The application driven centrifuge package ensures convenient, fast ordering of all components necessary to run a specific protocol. Run 16x50 mL conicals under biocontainment certified lids. Contains centrifuge, TX-400 rotor, buckets, biocontainment lids, 50 mL conical adapters (maximum capacity 16 x 50 mL), 15 mL conical adapter (maximum capacity 36 x 15 mL)
  • Easy to Use:
    • Intuitive controls simplify operation and provide detailed information on a wide range of operating parameters and processing status for optimal performance.
    • Modern industrial design supports continuous improvement in rotor loading ergonomics, user safety and current needed certifications
    • Motorized lid latch enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches.
    Thermo Scientific™ Auto-Lock™ III Rotor System:
    • Secure locking system allows easy push-button installation and exchange of rotors.
    • System saves time when switching between applications.
    • Easy access to rotor chamber offers quick cleaning, a healthier working environment and longer unit life.
    Thermo Scientific™ ClickSeal™ Bucket Sealing System:
    • Biocontainment solution seals with a snap; eliminates screw caps and complicated clips.
    • System has glove-friendly, one-handed open/close capability.
    • System is certified by CAMR™ in Porton Down, UK.
    Thermo Scientific™ SMARTSpin™ Technology:
    • Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option:
    • Technology optimizes safety and improves separations.
    Space-Saving Design:
    • Compact dimensions optimize work area and ergonomic height and curved design ensure easy sample loading and unloading and easy cleaning.
    Energy Efficient:
    • System offers up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Drive System Direct, Brushless induction low profile motor with choices of 9 acceleration, 10 deceleration rates
Includes Part Numbers In Pk. 75009261 (centrifuge),75003181(rotor),75003655(set of buckets),75003656(lids),75003682(set of adapters),75003683(set of adapters)
Product Type General Purpose Centrifuge Series