Learn More
Thermo Scientific™ Sorvall™ X1 R Pro, TX-400 Cell Culture Package PROMO
Catalog No. 75016012PM Shop All Thermo Scientific Products
Change view
Click to view available options
X1 R Pro, TX-400 Cell Culture Package; Includes: Centrifuge, Rotor, Set of Buckets, Lids, 2 Sets of Adapters
Catalog No.
75-016-012PM X1 R Pro, TX-400 Cell Culture Package; Includes: Centrifuge, Rotor, Set of Buckets, Lids, 2 Sets of Adapters
1 options
This item is not returnable. View return policy
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. 75-016-012PM Supplier Thermo Scientific™ Supplier No. 75016012PR
This item is not returnable. View return policy
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
This application-driven centrifuge package ensures convenient, fast ordering of all components necessary to run a specific protocol.
  • Intuitive controls simplify operation and provide detailed information on a wide range of operating parameters and processing status for optimal performance
  • Modern industrial design supports continuous improvement in rotor loading ergonomics, user safety and current needed certifications
  • Motorized lid latch enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches
  • Compact dimensions optimize work area and ergonomic height and curved design ensure easy sample loading and unloading and easy cleaning
    Thermo Scientific™ Auto-Lock™ III Rotor System:
    • Secure locking system allows easy push-button installation and exchange of rotors
    • System saves time when switching between applications
    • Easy access to rotor chamber offers quick cleaning, a healthier working environment and longer unit life
    Thermo Scientific™ ClickSeal™ Bucket Sealing System:
    • Biocontainment solution seals with a snap; eliminates screw caps and complicated clips
    • System has glove-friendly, one-handed open/close capability
    • System is certified by CAMR™ in Porton Down, UK
    Thermo Scientific™ SMARTSpin™ Technology:
    • Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option
    • Technology optimizes safety and improves separations
    Space-Saving Design:
  • Direct, Brushless Induction low profile motor with choices of 9 acceleration, 10 deceleration rates
  • Energy Efficient:
    • System offers up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing
TRUSTED_SUSTAINABILITY
Specifications
Drive System Direct, Brushless Induction Drive
Green Features Less Waste, Energy Efficient
Includes Centrifuge, Rotor, Set of Buckets, Lids, 2 Sets of Adapters
Voltage 120 V
Product Type General Purpose Centrifuge