Learn More
Medchemexpress LLC SOS1-IN-11 | 2654741-64-5 | 98.2% | C22H24F3N5O | 100 MG
SDP
Catalog No. 5000354831
Encompass_Preferred
SOS1-IN-11 is a potent SOS1 inhibitor with an IC50 value of 30 nM. It shows antiproliferative activity against human MKN-1 cells with an IC50 of 26 nM, as assessed by reduction in cell viability measured for 48 hours using a CCK-8 assay. It is for research use only and not intended for patient use.
  • Potent SOS1 inhibitor
  • IC50 value of 30 nM
  • Antiproliferative activity against human MKN-1 cells (IC50 of 26 nM)
  • Suitable for research use only
Catalog No. 50-003-54831 Supplier Medchemexpress LLC Supplier No. HY144962100MG
SOS1-IN-11 is a potent SOS1 inhibitor with an IC50 value of 30 nM. It shows antiproliferative activity against human MKN-1 cells with an IC50 of 26 nM, as assessed by reduction in cell viability measured for 48 hours using a CCK-8 assay. It is for research use only and not intended for patient use.
  • Potent SOS1 inhibitor
  • IC50 value of 30 nM
  • Antiproliferative activity against human MKN-1 cells (IC50 of 26 nM)
  • Suitable for research use only