Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SOS1-IN-11 | 2654741-64-5 | 98.2% | C22H24F3N5O | 10MG
SDP

Catalog No. 5000258536
Encompass_Preferred

SOS1-IN-11 is a small-molecule SOS1 inhibitor supplied for research use. It demonstrates potent biochemical inhibition of SOS1 and cellular antiproliferative activity in gastric cancer cells, and is provided as a light yellow solid with high listed purity.

  • Potent SOS1 inhibition (IC50 ≈ 30 nM).
  • Demonstrated antiproliferative activity in MKN-1 cells (IC50 ≈ 26 nM, 48 h, CCK-8 assay).
  • Molecular formula C22H24F3N5O and molecular weight 431.45 g·mol⁻¹.
  • High listed purity (98.22%).
  • Soluble in DMSO (100 mg/mL); sonication recommended for dissolution.
  • Powder is stable at -20°C for long-term storage.
  • Offered in multiple lab-scale pack sizes for research applications.

Catalog No. 50-002-58536 Supplier Medchemexpress LLC Supplier No. HY14496210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SOS1-IN-11 is a small-molecule SOS1 inhibitor supplied for research use. It demonstrates potent biochemical inhibition of SOS1 and cellular antiproliferative activity in gastric cancer cells, and is provided as a light yellow solid with high listed purity.

  • Potent SOS1 inhibition (IC50 ≈ 30 nM).
  • Demonstrated antiproliferative activity in MKN-1 cells (IC50 ≈ 26 nM, 48 h, CCK-8 assay).
  • Molecular formula C22H24F3N5O and molecular weight 431.45 g·mol⁻¹.
  • High listed purity (98.22%).
  • Soluble in DMSO (100 mg/mL); sonication recommended for dissolution.
  • Powder is stable at -20°C for long-term storage.
  • Offered in multiple lab-scale pack sizes for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.