Learn More
Medchemexpress LLC Sos1-in-11 | 2654741-64-5 | 98.2% | C22H24F3N5O | 50 MG
SDP
Catalog No. 5000355114
Encompass_Preferred
SOS1-IN-11 is a potent SOS1 inhibitor with an IC50 value of 30 nM. It demonstrates antiproliferative activity against human MKN-1 cells with an IC50 of 26 nM, assessed via CCK-8 assay over 48 hours. This product is for research use only.
  • Potent SOS1 inhibitor
  • IC50 of 30 nM against SOS1
  • Antiproliferative activity against human MKN-1 cells (IC50: 26 nM)
  • Suitable for research applications
Catalog No. 50-003-55114 Supplier Medchemexpress LLC Supplier No. HY14496250MG
SOS1-IN-11 is a potent SOS1 inhibitor with an IC50 value of 30 nM. It demonstrates antiproliferative activity against human MKN-1 cells with an IC50 of 26 nM, assessed via CCK-8 assay over 48 hours. This product is for research use only.
  • Potent SOS1 inhibitor
  • IC50 of 30 nM against SOS1
  • Antiproliferative activity against human MKN-1 cells (IC50: 26 nM)
  • Suitable for research applications