Learn More
ACROBiosystems ACROBiosystems Biotinylated Human SOST / Sclerostin Protein, His Tag, ultra sensitivity (primary amine labeling)
Catalog No. 501957534
Encompass_Preferred
Recombinant Protein;25UG;MABSol Biotinylated Human SOST, His Tag, primary amine labeling (SOT-H8245) is expressed from human HEK293 cells. It contains AA Gln 24 - Tyr 213 (Accession # AAI01087.1). It is the biotinylated form of Human SOST, His Tag (HST-H5245).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 22.3 kDa. The protein migrates as 25-36 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.
Catalog No. 50-195-7534 Supplier ACROBiosystems Supplier No. SOTH824525UG
Recombinant Protein;25UG;MABSol Biotinylated Human SOST, His Tag, primary amine labeling (SOT-H8245) is expressed from human HEK293 cells. It contains AA Gln 24 - Tyr 213 (Accession # AAI01087.1). It is the biotinylated form of Human SOST, His Tag (HST-H5245).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 22.3 kDa. The protein migrates as 25-36 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.