Learn More
Sino Biological Recombinant Human SOST/Sclerostin Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10593H07HB100
Encompass_Preferred
A DNA sequence encoding the Human SOST (NP_079513.1) (Gln24-Tyr213) was expressed with a polyhistidine tag at the N-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6229