Learn More
Medchemexpress LLC HY-15516 5mg Medchemexpress, Sotagliflozin CAS:1018899-04-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY155165MG
Encompass_Preferred
Medchemexpress, HY-15516 5mg Sotagliflozin CAS:1018899-04-1 Sotagliflozin (LX-4211) is a potent dual SGLT2/1 inhibitor. Antidiabetic agents. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1730