Learn More
Medchemexpress LLC Sotalol-d6 hydrochloride | 1246820-85-8 | 99.0% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0437AS5MG
Encompass_Preferred
Sotalol-d6 hydrochloride is a deuterium labeled Sotalol hydrochloride. It is an orally active, non-selective competitive β-adrenergic receptor blocker and a potent antiarrhythmic agent. It blocks β-receptors and potassium KCNH2 channels.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-08923