Learn More
Medchemexpress LLC Sotalol hydrochloride | 959-24-0 | 99.9% | 5 MG
SDP
Catalog No. 5000407868
Encompass_Preferred
Sotalol hydrochloride (MJ 1999) is an orally active, non-selective β-adrenergic receptor blocker. It is a potent antiarrhythmic agent used for the research of pediatric arrhythmias.
  • Orally active, non-selective β-adrenergic receptor blocker
  • Potent antiarrhythmic agent
  • Blocks β-receptors and potassium KCNH2 channels
  • Suitable for research on pediatric arrhythmias
  • Classified as an antiepileptic agent
Catalog No. 50-004-07868 Supplier Medchemexpress LLC Supplier No. HYB04375MG
Sotalol hydrochloride (MJ 1999) is an orally active, non-selective β-adrenergic receptor blocker. It is a potent antiarrhythmic agent used for the research of pediatric arrhythmias.
  • Orally active, non-selective β-adrenergic receptor blocker
  • Potent antiarrhythmic agent
  • Blocks β-receptors and potassium KCNH2 channels
  • Suitable for research on pediatric arrhythmias
  • Classified as an antiepileptic agent