Learn More
Medchemexpress LLC Sotalol hydrochloride | 959-24-0 | 99.9% | 500 MG
SDP
Catalog No. 5000407015
Encompass_Preferred
Sotalol hydrochloride is an orally active, non-selective β-adrenergic receptor blocker. It is a potent antiarrhythmic agent used for the research of pediatric arrhythmias. This compound blocks β-receptors and potassium KCNH2 channels, and acts as an antiepileptic agent.
  • Orally active
  • Non-selective β-adrenergic receptor blocker
  • Potent antiarrhythmic agent
  • Used for the research of pediatric arrhythmias
  • Blocks β-receptors
  • Blocks potassium KCNH2 channels
  • Acts as an antiepileptic agent
Catalog No. 50-004-07015 Supplier Medchemexpress LLC Supplier No. HYB0437500MG
Sotalol hydrochloride is an orally active, non-selective β-adrenergic receptor blocker. It is a potent antiarrhythmic agent used for the research of pediatric arrhythmias. This compound blocks β-receptors and potassium KCNH2 channels, and acts as an antiepileptic agent.
  • Orally active
  • Non-selective β-adrenergic receptor blocker
  • Potent antiarrhythmic agent
  • Used for the research of pediatric arrhythmias
  • Blocks β-receptors
  • Blocks potassium KCNH2 channels
  • Acts as an antiepileptic agent