Learn More
Medchemexpress LLC Sotalol hydrochloride | 959-24-0 | MFCD00242937 | 99.9% | 308.82 g/mol | C12H21ClN2O3S | 10 MG
SDP
Catalog No. 5000297996
Encompass_Preferred
Sotalol hydrochloride is an orally active, non-selective β-adrenergic receptor blocker with class III antiarrhythmic activity used in cardiovascular and electrophysiology research. It is supplied as a characterized research chemical with high purity and documented analytical data.
  • High purity as determined by LCMS (≈99.9%).
  • Molecular weight 308.82 g/mol and molecular formula C12H21ClN2O3S.
  • Used to study β-receptor blockade and cardiac repolarization (potassium channel) effects.
  • Supplied in small research pack sizes and solution formats (for example, 10 MG solid or 10 mM/1 mL solution).
Catalog No. 50-002-97996 Supplier Medchemexpress LLC Supplier No. HYB043710MG
Sotalol hydrochloride is an orally active, non-selective β-adrenergic receptor blocker with class III antiarrhythmic activity used in cardiovascular and electrophysiology research. It is supplied as a characterized research chemical with high purity and documented analytical data.
  • High purity as determined by LCMS (≈99.9%).
  • Molecular weight 308.82 g/mol and molecular formula C12H21ClN2O3S.
  • Used to study β-receptor blockade and cardiac repolarization (potassium channel) effects.
  • Supplied in small research pack sizes and solution formats (for example, 10 MG solid or 10 mM/1 mL solution).