Learn More
Medchemexpress LLC Sotalol hydrochloride | 959-24-0 | MFCD00870503 | ≥99.0% | 200 MG
SDP
Catalog No. 5000406779
Encompass_Preferred
Sotalol hydrochloride is an orally active, non-selective β-adrenergic receptor blocker and a potent antiarrhythmic agent. It is suitable for research on pediatric arrhythmias and also functions as an antiepileptic agent. This compound works by blocking β-receptors and potassium KCNH2 channels.
  • Orally active, non-selective β-adrenergic receptor blocker
  • Potent antiarrhythmic agent
  • Can be used for research of pediatric arrhythmias
  • Blocks β-receptors
  • Blocks potassium KCNH2 channels
  • Functions as an antiepileptic agent
  • Target for adrenergic receptor and potassium channel research
  • Relevant for cardiovascular disease and neurological disease research
  • Stable under recommended storage conditions
  • Available as a solid
Catalog No. 50-004-06779 Supplier Medchemexpress LLC Supplier No. HYB0437200MG
Sotalol hydrochloride is an orally active, non-selective β-adrenergic receptor blocker and a potent antiarrhythmic agent. It is suitable for research on pediatric arrhythmias and also functions as an antiepileptic agent. This compound works by blocking β-receptors and potassium KCNH2 channels.
  • Orally active, non-selective β-adrenergic receptor blocker
  • Potent antiarrhythmic agent
  • Can be used for research of pediatric arrhythmias
  • Blocks β-receptors
  • Blocks potassium KCNH2 channels
  • Functions as an antiepileptic agent
  • Target for adrenergic receptor and potassium channel research
  • Relevant for cardiovascular disease and neurological disease research
  • Stable under recommended storage conditions
  • Available as a solid