Learn More
Selleck Chemical LLC Sotalol HCl 50mg 959-24-0
SDP
Supplier:  Selleck Chemical LLC S250950MG
Encompass_Preferred
Sotalol is a non-selective beta blocker and a potassium channel blocker with an IC50 of 43 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-494-4