Learn More
Cayman Chemical Sotalol (hydrochloride), Purity: ≥98%, CAS: 959-24-0, 50 mg
SDP
Catalog No. 502973165
Encompass_Preferred
A β-AR antagonist (IC50s = 8.9 and 5.2 μM for β1- and β2-ARs, respectively) and a class III antiarrhythmic agent; decreases IK in guinea pig ventricular cells and prolongs action potential duration in electrically stimulated isolated guinea pig papillary muscles at 100 μM; decreases heart rate and increases blood pressure and the cardiac FRP in a canine model of PES-induced ventricular tachycardia
Catalog No. 50-297-3165 Supplier Cayman Chemical Supplier No. 1613650 MG
A β-AR antagonist (IC50s = 8.9 and 5.2 μM for β1- and β2-ARs, respectively) and a class III antiarrhythmic agent; decreases IK in guinea pig ventricular cells and prolongs action potential duration in electrically stimulated isolated guinea pig papillary muscles at 100 μM; decreases heart rate and increases blood pressure and the cardiac FRP in a canine model of PES-induced ventricular tachycardia