Learn More
TARGETMOL CHEMICALS INC SOTORASIB 10MG
SDP
Catalog No. 502576230
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sotorasib (AMG-510) is a selective and orally bioavailable KRAS G12C covalent inhibitor. purity: 99%
Catalog No. 50-257-6230 Supplier TARGETMOL CHEMICALS INC Supplier No. T868410MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sotorasib (AMG-510) is a selective and orally bioavailable KRAS G12C covalent inhibitor. purity: 99%