Learn More
Medchemexpress LLC Sotorasib-d7 | 2296729-00-3 | 98.9% | 567.64 | 10 MG
SDP
Catalog No. 5000436181
Encompass_Preferred
Sotorasib-d7 is a deuterium-labeled Sotorasib and a first-in-class, orally bioavailable, and selective KRAS G12C covalent inhibitor. It irreversibly inhibits KRAS G12C by locking it in an inactive GDP-bound state, leading to the regression of KRAS G12C-mutated locally advanced or metastatic non-small cell lung cancer (NSCLC).
  • Deuterium-labeled Sotorasib.
  • First-in-class, orally bioavailable, and selective KRAS G12C covalent inhibitor.
  • Irreversibly inhibits KRAS G12C by locking it in an inactive GDP-bound state.
  • Leads to the regression of KRAS G12C-mutated locally advanced or metastatic non-small cell lung cancer (NSCLC).
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-36181 Supplier Medchemexpress LLC Supplier No. HY114277S10MG
Sotorasib-d7 is a deuterium-labeled Sotorasib and a first-in-class, orally bioavailable, and selective KRAS G12C covalent inhibitor. It irreversibly inhibits KRAS G12C by locking it in an inactive GDP-bound state, leading to the regression of KRAS G12C-mutated locally advanced or metastatic non-small cell lung cancer (NSCLC).
  • Deuterium-labeled Sotorasib.
  • First-in-class, orally bioavailable, and selective KRAS G12C covalent inhibitor.
  • Irreversibly inhibits KRAS G12C by locking it in an inactive GDP-bound state.
  • Leads to the regression of KRAS G12C-mutated locally advanced or metastatic non-small cell lung cancer (NSCLC).
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.