Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sotorasib-d7 (AMG-510-d7) | 98.9% | 567.64 g/mol | C30H23D7F2N6O3 | 5 MG
SDP

Catalog No. 5000264367
Encompass_Preferred

Sotorasib-d7 (AMG-510-d7) is a deuterium-labeled internal standard of sotorasib intended for research and analytical use, suitable as a tracer or internal standard for quantitative NMR, GC-MS, and LC-MS workflows. Supplied as a high-purity solid, it is formulated for reliable analytical performance and stable storage under controlled conditions.

  • Deuterium-labeled internal standard suitable for NMR, GC-MS, and LC-MS.
  • High purity (98.9%) for consistent analytical results.
  • Molecular weight 567.64 g/mol and formula C30H23D7F2N6O3.
  • White to light yellow solid for straightforward handling.
  • Soluble in DMSO at ≥100 mg/mL for concentrated stock preparation.
  • Stable when stored at -20°C under nitrogen; in solvent: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-002-64367 Supplier Medchemexpress LLC Supplier No. HY114277S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sotorasib-d7 (AMG-510-d7) is a deuterium-labeled internal standard of sotorasib intended for research and analytical use, suitable as a tracer or internal standard for quantitative NMR, GC-MS, and LC-MS workflows. Supplied as a high-purity solid, it is formulated for reliable analytical performance and stable storage under controlled conditions.

  • Deuterium-labeled internal standard suitable for NMR, GC-MS, and LC-MS.
  • High purity (98.9%) for consistent analytical results.
  • Molecular weight 567.64 g/mol and formula C30H23D7F2N6O3.
  • White to light yellow solid for straightforward handling.
  • Soluble in DMSO at ≥100 mg/mL for concentrated stock preparation.
  • Stable when stored at -20°C under nitrogen; in solvent: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.