Learn More
Medchemexpress LLC Sotorasib racemate 100mg | 2252403-56-6 | 100 MG
SDP
Catalog No. 5000226450
Encompass_Preferred
Sotorasib racemate is the racemic form of sotorasib, a covalent KRAS G12C inhibitor for use in cancer research. Supplied as a research-grade solid, it includes reported purity, solubility, and storage recommendations for preclinical and in vitro studies.
  • Racemic sotorasib with covalent KRAS G12C inhibition
  • Suitable for in vitro and preclinical cancer research
  • Reported purity of 98.99%
  • Soluble in DMSO (≈3.85 mg/mL); ultrasonic agitation recommended
  • Store under nitrogen at -20°C; in solution store at -80°C for extended storage
Catalog No. 50-002-26450 Supplier Medchemexpress LLC Supplier No. HY114277A100MG
Sotorasib racemate is the racemic form of sotorasib, a covalent KRAS G12C inhibitor for use in cancer research. Supplied as a research-grade solid, it includes reported purity, solubility, and storage recommendations for preclinical and in vitro studies.
  • Racemic sotorasib with covalent KRAS G12C inhibition
  • Suitable for in vitro and preclinical cancer research
  • Reported purity of 98.99%
  • Soluble in DMSO (≈3.85 mg/mL); ultrasonic agitation recommended
  • Store under nitrogen at -20°C; in solution store at -80°C for extended storage