Learn More
Medchemexpress LLC HY-114277A 10mg , Sotorasib (racemate) CAS:2252403-56-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY114277A10MG
Encompass_Preferred
HY-114277A 10mg Sotorasib (racemate) CAS: 2252403-56-6 Sotorasib (AMG-510) racemate is the racemate of Sotorasib (AMG-510). AMG-510 is a potent, orally bioavailable, and selective KRAS G12C covalent inhibitor with anti-tumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0565