Learn More
Medchemexpress LLC Sotorasib racemate 50mg | 2252403-56-6 | 50 MG
SDP
Catalog No. 5000228846
Encompass_Preferred
Sotorasib racemate is the racemic form of sotorasib (AMG-510), a covalent KRAS G12C inhibitor used for preclinical research. Supplied as a solid powder in a 50 mg package suitable for formulation and in vitro testing.
  • Orally active KRAS G12C covalent inhibitor for research use.
  • Provided as a solid powder for assay preparation and formulation.
  • Suitable for cell-based and biochemical KRAS G12C studies.
  • High purity supports reproducible experimental results.
Catalog No. 50-002-28846 Supplier Medchemexpress LLC Supplier No. HY114277A50MG
Sotorasib racemate is the racemic form of sotorasib (AMG-510), a covalent KRAS G12C inhibitor used for preclinical research. Supplied as a solid powder in a 50 mg package suitable for formulation and in vitro testing.
  • Orally active KRAS G12C covalent inhibitor for research use.
  • Provided as a solid powder for assay preparation and formulation.
  • Suitable for cell-based and biochemical KRAS G12C studies.
  • High purity supports reproducible experimental results.