Learn More
Selleck Chemical LLC Sotrastaurin 5mg 425637-18-9 AEB071
SDP
Supplier:  Selleck Chemical LLC S27915MG
Encompass_Preferred
Sotrastaurin (AEB071) is a potent and selective pan-PKC inhibitor, mostly for PKC? with Ki of 0.22 nM in a cell-free assay; inactive to PKC?. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1075075