Learn More
Apexbio Technology LLC Sotrastaurin (AEB071)(Synonyms: AEB071, Sotrastaurin, PKC inhibitor AEB071, PKC inhibitor Sotrastaurin, Sotrastaurin base, AEB-071), 10mM (in 1mL DMSO), CAS: 425637-18-9.
SDP
Catalog No. 5000060108
Encompass_Preferred
Sotrastaurin (AEB071 CAS 425637-18-9) is a small-molecule inhibitor targeting protein kinase C (PKC) with inhibitory activity against classical and novel PKC isoforms PKC plays an integral role in immune cell signaling influencing T-cell activation proliferation and cytokine synthesis Sotrastaurin blocks PKC-mediated pathways thereby suppressing T-cell activation proliferation IL-2 mRNA expression and NK-cell function as demonstrated in ex vivo lymphocyte assays Due to its immunoregulatory properties Sotrastaurin has potential therapeutic applications in autoimmune conditions including psoriasis
Catalog No. 50-000-60108 Supplier Apexbio Technology LLC Supplier No. A85255.1
Sotrastaurin (AEB071 CAS 425637-18-9) is a small-molecule inhibitor targeting protein kinase C (PKC) with inhibitory activity against classical and novel PKC isoforms PKC plays an integral role in immune cell signaling influencing T-cell activation proliferation and cytokine synthesis Sotrastaurin blocks PKC-mediated pathways thereby suppressing T-cell activation proliferation IL-2 mRNA expression and NK-cell function as demonstrated in ex vivo lymphocyte assays Due to its immunoregulatory properties Sotrastaurin has potential therapeutic applications in autoimmune conditions including psoriasis