Learn More
Medchemexpress LLC Sotuletinib (hydrochloride) | 2222138-31-8 | 99.4% | 10 MG
SDP
Catalog No. 5000342127
Encompass_Preferred
Sotuletinib hydrochloride is a potent, selective, orally active, and brain-penetrant CSF-1R (c-Fms) inhibitor with an IC50 of 1 nM. It exhibits over 1,000-fold selectivity against its closest receptor tyrosine kinase homologs. This compound is used for microglia depletion and research related to tumor and CNS-related diseases.
  • Potent, selective, and orally active CSF-1R inhibitor
  • Brain-penetrant properties
  • Shows high selectivity against receptor tyrosine kinase homologs
  • Useful for microglia depletion
  • Applicable in tumor and CNS-related disease research
Catalog No. 50-003-42127 Supplier Medchemexpress LLC Supplier No. HY12768A10MG
Sotuletinib hydrochloride is a potent, selective, orally active, and brain-penetrant CSF-1R (c-Fms) inhibitor with an IC50 of 1 nM. It exhibits over 1,000-fold selectivity against its closest receptor tyrosine kinase homologs. This compound is used for microglia depletion and research related to tumor and CNS-related diseases.
  • Potent, selective, and orally active CSF-1R inhibitor
  • Brain-penetrant properties
  • Shows high selectivity against receptor tyrosine kinase homologs
  • Useful for microglia depletion
  • Applicable in tumor and CNS-related disease research