Learn More
Gilson Company Inc Sound Enclosure for W.S. Tyler Ro-Tap Sieve Shakers
Catalog No. NC1748754
Encompass_Preferred
SSA-320A Sound Enclosure for SS-30 or SS-31 W.S. Tyler Ro-Tap Sieve Shakers has a steel cabinet lined with 1in (25.4mm) thick acoustic foam to greatly reduce noise levels. Two front doors and a top-opening panel allow easy access. A small access port is built-in to the rear of each unit.
Catalog No. NC1748754 Supplier Gilson Company Inc Supplier No. SSA320A
SSA-320A Sound Enclosure for SS-30 or SS-31 W.S. Tyler Ro-Tap Sieve Shakers has a steel cabinet lined with 1in (25.4mm) thick acoustic foam to greatly reduce noise levels. Two front doors and a top-opening panel allow easy access. A small access port is built-in to the rear of each unit.