Learn More
Grainger REED INSTRUMENTS Digital Sound Level Meter Kit, 30 dB to 130 dB, Type 2, 1-Piece Set
Catalog No. 502761666
Encompass_Preferred
The sound level meter and calibrator kit is designed for precise acoustic measurements across a variety of industrial and environmental applications. This comprehensive kit includes a Type 2 sound level meter, a calibrator for field verification, and a professional-grade hard carrying case for secure transport and storage. The meter features a 4-digit backlit LCD and covers a wide measurement range of 30 dB to 130 dB with high accuracy.
  • High accuracy of 1.4 dB meeting Type 2 standards
  • Dual-range measurement with High and Low settings
  • A and C frequency weighting
  • Fast and Slow time weighting
  • Data hold and max hold functions
  • Tripod mountable for long-term monitoring
  • Backlit 4-digit LCD display
  • Low battery indicator and 15-minute auto shut-off
Catalog No. 50-276-1666 Supplier Grainger Supplier No. 161D41
The sound level meter and calibrator kit is designed for precise acoustic measurements across a variety of industrial and environmental applications. This comprehensive kit includes a Type 2 sound level meter, a calibrator for field verification, and a professional-grade hard carrying case for secure transport and storage. The meter features a 4-digit backlit LCD and covers a wide measurement range of 30 dB to 130 dB with high accuracy.
  • High accuracy of 1.4 dB meeting Type 2 standards
  • Dual-range measurement with High and Low settings
  • A and C frequency weighting
  • Fast and Slow time weighting
  • Data hold and max hold functions
  • Tripod mountable for long-term monitoring
  • Backlit 4-digit LCD display
  • Low battery indicator and 15-minute auto shut-off