Learn More
Grainger REED Instruments Sound Level Meter, Datalogger, 30dB To 130dB, Type 2, 1 ea
Catalog No. 502761682
Encompass_Preferred
This sound level meter and datalogger is designed for accurate sound pressure level measurements in applications such as preventative maintenance. It offers wide measurement ranges and internal data logging capabilities to monitor noise levels effectively.
  • Measures from 30 dB to 130 dB with 1.4 dB accuracy.
  • Includes 1 MB of internal memory to record datapoints with time and date stamps.
  • Features A and C frequency weighting and fast/slow time weighting.
  • Equipped with a 0.5 in. (12.7 mm) electret condenser microphone.
  • Large backlit 4-digit LCD display with a 30-segment digital analog bargraph.
  • User-selectable sampling rate from 1 sec. to 60 sec.
  • PC connectivity via USB.
  • Includes windshield, USB cable, PC software, hard carrying case and batteries.
  • Meets IEC 61672-1 Class 2 and ANSI S1.4 Type 2 standards.
  • Tripod mountable.
Catalog No. 50-276-1682 Supplier Grainger Supplier No. 161D43
This sound level meter and datalogger is designed for accurate sound pressure level measurements in applications such as preventative maintenance. It offers wide measurement ranges and internal data logging capabilities to monitor noise levels effectively.
  • Measures from 30 dB to 130 dB with 1.4 dB accuracy.
  • Includes 1 MB of internal memory to record datapoints with time and date stamps.
  • Features A and C frequency weighting and fast/slow time weighting.
  • Equipped with a 0.5 in. (12.7 mm) electret condenser microphone.
  • Large backlit 4-digit LCD display with a 30-segment digital analog bargraph.
  • User-selectable sampling rate from 1 sec. to 60 sec.
  • PC connectivity via USB.
  • Includes windshield, USB cable, PC software, hard carrying case and batteries.
  • Meets IEC 61672-1 Class 2 and ANSI S1.4 Type 2 standards.
  • Tripod mountable.