Learn More
Grainger PORTA-KING Sound Reducing Equipment Enclosure, 10 ft x 12 ft x 8 ft, Steel-Covered Drywall, STC 34, 1 ea
Catalog No. 502777746
Encompass_Preferred
This sound-reducing equipment enclosure is engineered to mitigate noise pollution from industrial machinery such as pumps, compressors, and generators. Designed for on-site assembly, this unit features a durable steel-covered drywall construction with an STC 34 sound transmission rating, making it suitable for maintaining workplace noise compliance in manufacturing and commercial environments.
  • Provides 95 sq ft of interior workspace
  • Features Class A fire rating
  • Includes comprehensive electrical package with 100 A circuit breaker panel, three light fixtures, two 110 V AC electrical outlets, and one switch
  • Constructed with R-9 wall insulation and R-19 ceiling insulation
  • Equipped with a non-combustible lay-in acoustical ceiling system
  • Includes one hinged door with lockset and tempered safety glass
Catalog No. 50-277-7746 Supplier Grainger Supplier No. 18C737
Add to Cart
Add to Cart
Request a Quote
This sound-reducing equipment enclosure is engineered to mitigate noise pollution from industrial machinery such as pumps, compressors, and generators. Designed for on-site assembly, this unit features a durable steel-covered drywall construction with an STC 34 sound transmission rating, making it suitable for maintaining workplace noise compliance in manufacturing and commercial environments.
  • Provides 95 sq ft of interior workspace
  • Features Class A fire rating
  • Includes comprehensive electrical package with 100 A circuit breaker panel, three light fixtures, two 110 V AC electrical outlets, and one switch
  • Constructed with R-9 wall insulation and R-19 ceiling insulation
  • Equipped with a non-combustible lay-in acoustical ceiling system
  • Includes one hinged door with lockset and tempered safety glass