Learn More
Grainger Porta-King Sound Reducing Equipment Enclosure, 8 ft x 8 ft x 8 ft, Steel-Covered Drywall, STC 34
Catalog No. 502777692
Encompass_Preferred
This sound-reducing equipment enclosure provides a controlled environment for industrial noise management applications. Engineered with a durable steel-covered drywall construction, this modular unit features a sound transmission coefficient of 34 STC to effectively mitigate noise levels within the facility. The enclosure includes a comprehensive electrical package with outlets and lighting for immediate operational readiness.
  • Sound transmission coefficient of 34 STC
  • Constructed with steel-covered drywall panels
  • Equipped with an electrical package including two 110 V AC outlets
  • Includes one T8 light fixture and one electrical switch
  • Features a non-combustible lay-in acoustical ceiling system
  • Clear satin anodized aluminum frame
  • Hinged aluminum door with included lockset
  • Includes 100 A circuit breaker panel
  • Class A fire rating
Catalog No. 50-277-7692 Supplier Grainger Supplier No. 18C735
Add to Cart
Add to Cart
Request a Quote
This sound-reducing equipment enclosure provides a controlled environment for industrial noise management applications. Engineered with a durable steel-covered drywall construction, this modular unit features a sound transmission coefficient of 34 STC to effectively mitigate noise levels within the facility. The enclosure includes a comprehensive electrical package with outlets and lighting for immediate operational readiness.
  • Sound transmission coefficient of 34 STC
  • Constructed with steel-covered drywall panels
  • Equipped with an electrical package including two 110 V AC outlets
  • Includes one T8 light fixture and one electrical switch
  • Features a non-combustible lay-in acoustical ceiling system
  • Clear satin anodized aluminum frame
  • Hinged aluminum door with included lockset
  • Includes 100 A circuit breaker panel
  • Class A fire rating