Learn More
Grainger Porta-King Sound Reducing Equipment Enclosure, Steel-Covered Drywall, 8 ft x 8 ft x 12 ft, 1 ea
Catalog No. 502776062
Encompass_Preferred
This sound reducing equipment enclosure provides a controlled environment for machinery and equipment. Constructed with durable steel-covered drywall walls, it is designed to minimize noise transmission while offering a secure and functional workspace.
  • Sound transmission coefficient of STC 34
  • Class A fire rating for enhanced safety
  • Includes electrical package with 110 V AC outlets
  • Equipped with two light fixtures for T8 light bulbs
  • Features a hinged door made of 20 ga aluminum
  • Ceiling system includes a non-combustible lay-in acoustical design with R-19 insulation
  • Includes lockset and mounting hardware for installation
  • Shipped unassembled
Catalog No. 50-277-6062 Supplier Grainger Supplier No. 18C736
Add to Cart
Add to Cart
Request a Quote
This sound reducing equipment enclosure provides a controlled environment for machinery and equipment. Constructed with durable steel-covered drywall walls, it is designed to minimize noise transmission while offering a secure and functional workspace.
  • Sound transmission coefficient of STC 34
  • Class A fire rating for enhanced safety
  • Includes electrical package with 110 V AC outlets
  • Equipped with two light fixtures for T8 light bulbs
  • Features a hinged door made of 20 ga aluminum
  • Ceiling system includes a non-combustible lay-in acoustical design with R-19 insulation
  • Includes lockset and mounting hardware for installation
  • Shipped unassembled