Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Sound Resonance Kit
GREENER_CHOICESCI_ED

Catalog No. S81865 Shop All Supertek Scientific Products
Change view
Click to view available options
Frequency:
256 Hz
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frequency
S81865 256 Hz
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S81865 Supplier Supertek™ Supplier No. PH36141S3
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fun hands-on experiments with Supertek™ Sound Resonance Kit. The pair of tuning forks demonstrate the resonance of sound and other acoustic phenomena. Kit includes two identical tuning forks, a rubber striker, a piece of clay to vary the frequency and a plastic dish for sound wave observation.

  • Kit includes:
    • 2x 256 Hz 'C' tuning forks
    • 1x Rubber striker with handle
    • 1x Piece of clay
    • 1x 70 mm Plastic dish

Specifications

Product Type Tuning Fork
For Use With (Application) Science Experiments
Includes 2 Tuning Forks, a Striker, a Piece of Clay, and a Plastic Dish
Diameter (Metric) 70 mm Dish
Frequency 256 Hz
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.