Learn More
Thermo Scientific SOURCE LC CONNECTION KIT
Catalog No. NC2282892 Shop All Thermo Scientific Products
Encompass
Fitting, fingertight, two-piece, one wing, 10-32 (Upchurch Scientific™ F-200, 2 provided) Grounding union, zero-dead-volume (ZDV), stainless steel, 1/16 in. orifice, 0.010 in. (0.25 mm) thru-hole, 10-32 (Upchurch Scientific U-435) Tubing, natural PEEK, 1/16 in. OD, 0.0025 in. ID, 28 cm (11 in.) long Tubing, red PEEK, 1/16 in. OD, 0.005 in. ID, 18 cm (7.1 in.) long (2 provided)
This item is not returnable. View return policy
Catalog No. NC2282892 Supplier Thermo Scientific Supplier No. 8000062057
Fitting, fingertight, two-piece, one wing, 10-32 (Upchurch Scientific™ F-200, 2 provided) Grounding union, zero-dead-volume (ZDV), stainless steel, 1/16 in. orifice, 0.010 in. (0.25 mm) thru-hole, 10-32 (Upchurch Scientific U-435) Tubing, natural PEEK, 1/16 in. OD, 0.0025 in. ID, 28 cm (11 in.) long Tubing, red PEEK, 1/16 in. OD, 0.005 in. ID, 18 cm (7.1 in.) long (2 provided)