Learn More
Perkin Elmer US LLC Source Lens #3 for SMARTsource
Catalog No. 502117646
Encompass_Preferred
For use with EI/CI ions in SMARTsource (Simplified Maintenance And Removal Technology).
Catalog No. 50-211-7646 Supplier Perkin Elmer US LLC Supplier No. N6480149
For use with EI/CI ions in SMARTsource (Simplified Maintenance And Removal Technology).