Learn More
Waters Corp Waters Corporation Test Kit, Mass Spectrometry, Source Pressure, Performance Maintenance
Catalog No. 50466753 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit is designed for conducting source pressure tests on mass spectrometry instruments, particularly those with serial numbers E06EMD058P and later. It is an essential component of the Performance Maintenance Protocol, ensuring the optimal functionality and reliability of mass spectrometry systems.
  • Used for source pressure testing.
  • Supports performance maintenance protocols.
  • Ensures optimum performance of mass spectrometry systems.
  • Aids in trouble-free operation of instruments.
  • Applicable for specific instrument series (E06EMD058P onwards).
Catalog No. 50-466-753 Supplier Waters Corp Supplier No. S100981DC1
This kit is designed for conducting source pressure tests on mass spectrometry instruments, particularly those with serial numbers E06EMD058P and later. It is an essential component of the Performance Maintenance Protocol, ensuring the optimal functionality and reliability of mass spectrometry systems.
  • Used for source pressure testing.
  • Supports performance maintenance protocols.
  • Ensures optimum performance of mass spectrometry systems.
  • Aids in trouble-free operation of instruments.
  • Applicable for specific instrument series (E06EMD058P onwards).