Learn More
ERA WATERS CORP SOURCE WATER E-COLI
SDP
Supplier:  ERA WATERS CORP 078
Encompass
Each sample is one lyophilized quantitative standard containing E. coli in the range 20-200 CFU/100mL or MPN/100mL. Use with all SDWA quantitative methods. Each standard can be used for Total Coliform, Fecal Coliform and E. coli.
Catalog No. NC0052979