Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SouthernBiotech™ Normal Rabbit Sera
SDP

Catalog No. OB004001 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB004001 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Reagents contain sodium azide. Sodium azide is very toxic if ingested or inhaled. Avoid contact with skin, eyes, or clothing. Wear eye or face protection when handling. If skin or eye contact occurs, wash with copious amounts of water. If ingested or inhaled, contact a physician immediately. Sodium azide yields toxic hydrazoic acid under acidic conditions. Dilute azide-containing compounds in running water before discarding to avoid accumulation of potentially explosive deposits in lead or copper plumbing.
Catalog No. OB004001 Supplier SouthernBiotech™ Supplier No. 004001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reagents contain sodium azide. Sodium azide is very toxic if ingested or inhaled. Avoid contact with skin, eyes, or clothing. Wear eye or face protection when handling. If skin or eye contact occurs, wash with copious amounts of water. If ingested or inhaled, contact a physician immediately. Sodium azide yields toxic hydrazoic acid under acidic conditions. Dilute azide-containing compounds in running water before discarding to avoid accumulation of potentially explosive deposits in lead or copper plumbing.

Processed to prevent minimal hemolysis and filtered to 0.2μm

Blocking sera; Control sera; Serum additive in dilution buffers

Specifications

Certifications/Compliance RUO
Color Colorless to Light Brown
Content And Storage -20°C
Species Adult New Zealand Rabbits
Form Liquid
Product Type Rabbit Serum
Quantity 5 mL
For Use With (Application) Blocking Sera, control Sera, Serum additive in dilution Buffers
Isotype IgG
pH 8
Preservative 0.1% NaN3
Show More Show Less

For Research Use Only. Not for Diagnostic or Therapeutic Use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.