Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Southern Blot Analysis For DNA Fingerprinting
SDPSCI_ED

Catalog No. S11207 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
5 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S11207 5 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S11207 Supplier Edvotek™ Supplier No. 207
Add to Cart
Add to Cart

Includes

instructions, DNA samples for electrophoresis, practice gel loading solution, UltraSpec-Agarose™, electrophoresis buffer, pipets, 5 pre-cut nylon membranes, 5 pre-cut blotting filter papers, Blue-Blot DNA Stain™

Requires

  • Horizontal gel electrophoresis apparatus
  • DC power supply
  • Waterbath (65°C)
  • Visualization system (white light)
  • Staining tray and net
  • NaCl, NaOH, concentrated HCl
  • 250mL beakers or flasks
  • 10mL glass graduated cylinder
  • 100mL graduated cylinder
  • Distilled or deionized water
  • Microwave, hot plate, or Bunsen burner
  • Small plastic box or tray forceps

Advanced-level experiment for DNA fingerprinting.

This experiment introduces your students to Southern blotting as a tool for 'DNA Fingerprinting' in a hypothetical paternity determination. DNA fragments are first separated by agarose gel electrophoresis, then transferred to a nylon membrane and finally visualized by staining.

Specifications

Grade 9 to Undergraduate
For Use With (Application) Southern blotting
Includes Instructions, DNA samples for electrophoresis, Practice gel loading solution, UltraSpec-Agarose™, Electrophoresis buffer, Pipets, 5 pre-cut nylon membranes, 5 pre-cut blotting filter papers, Blue-Blot DNA Stain™
Requires Electrophoresis apparatus, Power supply, 65°C Water bath, DNA visualization system, Staining net and tray, Automatic micropipettes, Lab glassware, Microwave or hot plate, Distilled water, NaCl, NaOH, Concentrated HCl, Plastic wrap, Forceps
Time Range Electrophoresis: 45 minutes; Blot: overnight; Stain/Destain: 10 minutes
Class Size 5 Lab Groups
Product Type Electrophoresis Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.