Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SouthernBiotech™ Dapi-Fluoromount-G™ Clear Mounting Media
SDP

Catalog No. OB010020 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Volume (Metric):
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
OB010020 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB010020 Supplier SouthernBiotech™ Supplier No. 010020
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dapi-Fluoromount-G is a water-soluble, instant-blue nuclear probe fluorescing (455nm) compound for mounting fixed slides using a staining procedure having an aqueous final step. Mounting slides with Dapi-Fluoromount-G stains the cell nucleus and provides a semi-permanent seal for long-term storage of slide preparations.

  • Prepare cytocentrifuge preps according to established procedures
  • Fix and rehydrate preps
  • Remove one slide at a time from aqueous buffer, add 1 drop of Dapi-Fluoromount-G directly to cell prep, mount cover slip and press gently with a gauze sponge to remove excess mounting medium and to seal the cover slip
  • Allow mounted preps to air dry for 5 minutes before examination

Slide Mounting

Specifications

Volume (English) 0.67 oz.
Content And Storage Room Temperature
Volume (Metric) 20 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.