Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG(H+L) Donkey anti-Rat, Mouse SP ads, FITC, Polyclonal, Southern Biotech™
SDP

Catalog No. OB643002 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
1.0 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB6430-02 1.0 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB6430-02 Supplier Southern Biotechnology Associates Supplier No. 643002
Add to Cart
Add to Cart

Donkey Polyclonal Secondary Antibody

SouthernBiotech is a leading provider of polyclonal secondary antibodies offering high sensitivity and low background for use in a variety of applications.

Specifications

Antigen IgG(H+L)
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate FITC
Formulation Supplied in PBS/NaN3
Host Species Donkey
Immunogen Rat IgG
Purification Method Affinity Purified
Quantity 1.0 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rat
Content And Storage Store at 2-8°C; Protect from light
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.