Learn More
TARGETMOL CHEMICALS INC SOVAPREVIR 5MG
SDP
Catalog No. 502580489
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sovaprevir is a non-structural 3 (NS3) protease inhibitor with antiviral activity for the treatment of HCV infection. purity: 98%
Catalog No. 50-258-0489 Supplier TARGETMOL CHEMICALS INC Supplier No. T288305MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sovaprevir is a non-structural 3 (NS3) protease inhibitor with antiviral activity for the treatment of HCV infection. purity: 98%