Learn More
Medchemexpress LLC Sovesudil hydrochloride | 2984963-50-8 | 99.5% | 443.90 | 100 MG
SDP
Catalog No. 5000345262
Encompass_Preferred
Sovesudil hydrochloride is a potent, ATP-competitive Rho kinase (ROCK) inhibitor. It effectively lowers intraocular pressure (IOP) without causing hyperemia. This product is provided as a solution in DMSO.
  • Potent ATP-competitive Rho kinase inhibitor
  • Lowers intraocular pressure
  • Does not induce hyperemia
Catalog No. 50-003-45262 Supplier Medchemexpress LLC Supplier No. HY109191A10MM/1ML
Sovesudil hydrochloride is a potent, ATP-competitive Rho kinase (ROCK) inhibitor. It effectively lowers intraocular pressure (IOP) without causing hyperemia. This product is provided as a solution in DMSO.
  • Potent ATP-competitive Rho kinase inhibitor
  • Lowers intraocular pressure
  • Does not induce hyperemia