Learn More
Medchemexpress LLC Sovesudil hydrochloride | 2984963-50-8 | 99.5% | 443.90 | 25 MG
SDP
Catalog No. 5000347053
Encompass_Preferred
Sovesudil (PHP-201) hydrochloride is a potent, ATP-competitive, locally acting Rho kinase (ROCK) inhibitor with IC50s of 3.7 and 2.3 nM for ROCK-I and ROCK-II, respectively. It lowers intraocular pressure (IOP) without inducing hyperemia.
  • Potent, ATP-competitive, locally acting Rho kinase (ROCK) inhibitor
  • IC50s of 3.7 nM for ROCK-I and 2.3 nM for ROCK-II
  • Lowers intraocular pressure (IOP) without inducing hyperemia
  • Induces altered cellular behavior of human trabecular meshwork (HTM) cells in vitro
  • Effectively reduces IOP in ocular normotensive and acute hypertensive rabbits without causing distinct hyperemia in vivo
Catalog No. 50-003-47053 Supplier Medchemexpress LLC Supplier No. 25MGHY109191A
Sovesudil (PHP-201) hydrochloride is a potent, ATP-competitive, locally acting Rho kinase (ROCK) inhibitor with IC50s of 3.7 and 2.3 nM for ROCK-I and ROCK-II, respectively. It lowers intraocular pressure (IOP) without inducing hyperemia.
  • Potent, ATP-competitive, locally acting Rho kinase (ROCK) inhibitor
  • IC50s of 3.7 nM for ROCK-I and 2.3 nM for ROCK-II
  • Lowers intraocular pressure (IOP) without inducing hyperemia
  • Induces altered cellular behavior of human trabecular meshwork (HTM) cells in vitro
  • Effectively reduces IOP in ocular normotensive and acute hypertensive rabbits without causing distinct hyperemia in vivo