Learn More
Medchemexpress LLC HY-145598 5mg , Sovleplenib CAS:1415792-84-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1455985MG
Encompass_Preferred
HY-145598 5mg Sovleplenib CAS: 1415792-84-5 Sovleplenib (HMPL-523) is a highly potent, orally available and selective SYK inhibitor with an IC50 of 25 nM. Anti-tumor activity. Sovleplenib can be used for the research of immune thrombocytopenia (ITP). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1474