Learn More
Aviva Systems Biology Corp SOX1 Antibody - middle region (ARP38720_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38720_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SOX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SOX1. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Zebrafish
Catalog No. 50-014-56768