Learn More
Diagnostic Biosystems SOX10 SOX10 (Melanoma Marker)
Catalog No. 502407522
Encompass_Preferred
SOX10 is widely expressed in normal human tissues including melanocytes and breast tissue. SOX10 is also an important marker in malignant tumors such as melanoma, breast carcinoma, gliomas, and benign tumors such as schwannomas.SOX-10 is considered as a very reliable marker for recognizing residual desmoplastic melanomas. In normal tissues, it is expressed in Schwann cells, melanocytes and myoepithelial cells ofsalivary, bronchial and mammary glands. SOX-10 expression is also observed in mast cells.
Catalog No. 50-240-7522 Supplier Diagnostic Biosystems Supplier No. PDM565
SOX10 is widely expressed in normal human tissues including melanocytes and breast tissue. SOX10 is also an important marker in malignant tumors such as melanoma, breast carcinoma, gliomas, and benign tumors such as schwannomas.SOX-10 is considered as a very reliable marker for recognizing residual desmoplastic melanomas. In normal tissues, it is expressed in Schwann cells, melanocytes and myoepithelial cells ofsalivary, bronchial and mammary glands. SOX-10 expression is also observed in mast cells.