Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 6663MSM3XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: SPM607. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b kappa. Positive Control: HepG2 cells. Melanomas, breast carcinomas, gliomas. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant fragment (aa 115-269) of human SOX10 protein with hexa-histidine tag. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 49-58kDa.Chromosome: 22q13.1. Gene Symbol: SOX10. Gene ID: 6663. Ensemble: ENSG00000100146. Uniprot: NP_008872.
Catalog No. 50-188-5183