Learn More
Biocare Medical, LLC SOX10 Antibody - Concentrate, 1.0 ml
SDP
Supplier:  Biocare Medical, LLC ACI3099C
Encompass_Preferred
The SOX10 protein is widely expressed in normal human tissues including melanocytes and breast tissue. It is also an important marker in malignant tumors such as melanoma, breast carcinoma, gliomas and benign tumors such as schwannomas. SOX10 antibody has been shown to be expressed in 97-100% of desmoplastic and spindle cell melanomas and was also shown to be expressed in 100% of nevi. The majority of oligodendrogliomas but also a large percentage of astrocytomas and poorly differentiated glioblastomas have also been shown to express SOX10. PATENT PENDING.[message_box bg="#005581"]
Archives - New Ms Mono SOX10 Paper [button text='Download' link='http://www.archivesofpathology.org/doi/pdf/10.5858/arpa.2014-0077-OA' style='small alt-button white' target='blank']
[/message_box]
Catalog No. 50-719-733